Capping Reagens 80/10/10 THF/Pyridine/Acetic Anhydride (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™

Lieferant: Honeywell Chemicals
Gefahr

BR641-4X2.5L
HONCBR641-4X2.5LEA 3071.44 EUR
HONCBR641-4X2.5L
Capping Reagens 80/10/10 THF/Pyridine/Acetic Anhydride (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Capping Reagens

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis.


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.

Siedepunkt: 66 °C (main component)
Schmelzpunkt: -108,5 °C (main component)
Dichte: 0,888 g/cm³ (20 °C) (Hauptbestandteil)
Flammpunkt: -15 °C
Lagertemperatur: Raumtemperatur
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR