Capping Reagens 84/16 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Lieferant: Honeywell Chemicals
|
Gefahr
|
BR650-4X2.5L
HONCBR650-4X2.5LEA
1254.59
EUR
HONCBR650-4X2.5L
Capping Reagens 84/16 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ zur DNA/RNA Synthese, Burdick & Jackson™
Capping Reagens
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
UNSPSC Code: 12350000
Dichte:
0,910 g/cm3 (25 °C) Flammpunkt: –15 °C Lagertemperatur: Raumtemperatur |
UN:
2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...