Sulphur ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1178-100ML
HONCE1178-100MLEA 134.39 EUR
HONCE1178-100ML
Sulphur ICP standards, (CRMs), Fluka™
Standards Kationischer Standards Schwefel-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).