Yttrium ICP Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1196-100ML
HONCE1196-100MLEA 141.87 EUR
HONCE1196-100ML
Yttrium ICP Standards, (CRMs), Fluka™
Standards Kationischer Standards Yttrium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).