Yttrium ICP Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1196-100ML
HONCE1196-100MLEA
141.87
EUR
HONCE1196-100ML
Yttrium ICP Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Yttrium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).