Germanium ICP & ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1321-100ML
HONCE1321-100MLEA
110.48
EUR
HONCE1321-100ML
Germanium ICP & ICP-MS Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Germanium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).