Rhenium ICP and ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1344-100ML
HONCE1344-100MLEA
204.63
EUR
HONCE1344-100ML
Rhenium ICP and ICP-MS Standards, (CRMs), Fluka™
Standards
Kationischer Standards
Rhenium-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).