Aluminium ICP & ICP-MS Standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1370-100ML
HONCE1370-100MLEA 94.03 EUR
HONCE1370-100ML
Aluminium ICP & ICP-MS Standards, (CRMs), Fluka™
Standards Kationischer Standards Aluminium-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).