Arsenic(III) ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1500-100ML
HONCE1500-100MLEA
120.29
EUR
HONCE1500-100ML
Arsenic(III) ICP standards, (CRMs), Fluka™
Standards
Kationischer Standards
Arsen-Standards
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).