Arsenic(III) ICP standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E1500-100ML
HONCE1500-100MLEA 120.29 EUR
HONCE1500-100ML
Arsenic(III) ICP standards, (CRMs), Fluka™
Standards Kationischer Standards Arsen-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).