Phosphorus AAS standards, (CRMs), Fluka™
Lieferant: Honeywell Chemicals
E4114-100ML
HONCE4114-100MLEA 80.2 EUR
HONCE4114-100ML
Phosphorus AAS standards, (CRMs), Fluka™
Standards Kationischer Standards Phosphor-Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Standards: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).