Ammoniumfluorid

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Fluoroammonium

01105.9025
HONE01105.9025EA 0 EUR
HONE01105.9025
Ammoniumfluorid
Ammonium fluoride
Formel: NH₄F
Molecular Weight: 37,04 g/mol
Schmelzpunkt: 160 °C
Dichte: 1,0088…1.0096 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR