Bariumnitrat
Lieferant: Honeywell Chemicals
|
Gefahr
|
11420.9025
HONE11420.9025EA
0
EUR
HONE11420.9025
Bariumnitrat
Barium nitrate
Formel:
Ba(NO₃)₂ Molecular Weight: 261,34 g/mol Schmelzpunkt: ∼592 °C Dichte: ∼3,23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003442 CAS-Nummer: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II Merck Index: 13,00986 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...