Eisen(III)sulfat

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Dieisentrissulfat

12357.9020
HONE12357.9020EA 0 EUR
HONE12357.9020
Eisen(III)sulfat
Eisen(III)sulfat
Formel: Fe₂O₁₂S₃
Molecular Weight: 399,88 g/mol
Dichte: 3,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011007
CAS-Nummer: 10028-22-5
EINECS: 233-072-9
Merck Index: 12,04079

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR