Ammoniumformiat, rein

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Formic acid ammonium salt

25204.9030
HONE25204.9030EA 0 EUR
HONE25204.9030
Ammoniumformiat, rein
Ammonium formate
Formel: HCO₂NH₄
Molecular Weight: 63,06 g/mol
Schmelzpunkt: 119…121 °C
Dichte: 1,28 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013103
CAS-Nummer: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR