Snooded Polypropylene Caps
Lieferant: KOLMI HOPEN
70.401
HOPE70.401EA 118 EUR
HOPE70.401
Snooded Polypropylene Caps
Einweg-Kleidung
These snooded caps provide extra protection for workers with long hair in industrial environments.
  • Fully protects and contains long hair
  • Conveys professional image
  • Ability to use color code for tracking
  • Single use
Lieferumfang: Includes caps and dispenser box.