Solvent S-316 for oil extraction

Lieferant: HORIBA

Synonyme: FLON

5200100690
HORI5200100690EA 1060 EUR
HORI5200100690
Solvent S-316 for oil extraction
Solvent S-316 for oil extraction
CAS-Nummer: 9002-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR