Formalinlösung-Lösung 10% Fixativ für die Histologie, neutral gepuffert
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Formaldehyd, Methanal, Formol, Methylaldehyd
M-Clarity™ Qualitätsstufe = MQ200
HT501850-200L
HT501850-200LEA
953
EUR
HT501850-200L
Formalinlösung-Lösung 10% Fixativ für die Histologie, neutral gepuffert
Formalinlösung
Formalin solution, neutral buffered, 10% may be used as a reagent for the primary fixation of tissues for diagnostic applications of an immunoperoxidase method in hematopathology.
Formel:
CH₂O Molecular Weight: 30,03 g/mol Dichte: 1,080 g/cm3 Lagertemperatur: Room Temperature |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 |
Spezifikation Testergebnisse
Appearance (Colour) | Colorless |
Appearance (Turbidity) | Clear |
Performance | Pass |
Fixation test within acceptable limits | |
pH | 6.90 - 7.10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...