Gateway, Profibus
Lieferant: HUBER
10503
HUBE10503EA 0 EUR
HUBE10503
Gateway, Profibus
Regler Temperaturregler
This gateway enables the connection of temperature control units to Profibus systems.
Offers a range of possibilities for data communication within SPS and process control systems.
Bestellinformation: Includes a mounting bracket and connection cable.