Cryopreservation media, HyClone™ HyCryo-STEM™
Lieferant: HyClone products (Cytiva)
SR30002.02
HYCLSR30002.02EA
92.7
EUR
HYCLSR30002.02
Cryopreservation media, HyClone™ HyCryo-STEM™
Zelleinfriermedien
HyClone™ HyCryo-STEM is a serum-free cryopreservation media designed specifically to maintain differentiation potential and minimise spontaneous differentiation of stem cells.
- Greater recovery of viable neural progenitor stem cells compared to industry standard cryopreservation media (growth media containing 10% DMSO)
- Achieves similar recovery of viable human stem cells compared to the industry standard cryopreservation media (a cocktail of 25% serum replacement and 10% DMSO)
Note: For further manufacturing or research use. Not for diagnostic or therapeutic use.
Lieferumfang: Supplied as a 2X solution, 0,2 µm sterile filtered.