Guard column holder for DNAPac™ RP columns
Lieferant: Thermo Fisher Scientific
950-00
HYPE950-00EA 603 EUR
HYPE950-00
Guard column holder for DNAPac™ RP columns
Vorsäulenhalter
  • Column Format: Preparative Column, Standard
  • Diameter (Metric): 21,2 mm
  • Length (Metric): 360 mm
  • Temperature: Up to 110 °C