Ecolite(+)™ liquid scintillation cocktail, MP Biomedicals
Lieferant: MP Biomedicals
0188247501
ICNA0188247501EA 307 EUR
ICNA0188247501
Ecolite(+)™ liquid scintillation cocktail, MP Biomedicals
Szintillationsgemische
EcoLite(+)™ is a unique formulation of media possessing exceptional water and salt holding characteristics. Ready-to-use scintillation cocktail for a wide variety of aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is essential.
  • Environmentally safe
  • Non toxic and non flammable
  • Ready-to-use
  • Versatile
  • Excellent stability for RIA and receptor assays
  • Forms no gel or phasing
  • Ideal for all types of samples
Suitable for difficult-to-count RIA samples, basic samples, phosphate buffer samples, and biological fluids.