Ecolite(+)™ liquid scintillation cocktail, MP Biomedicals
Lieferant: MP Biomedicals
0188247501
ICNA0188247501EA
307
EUR
ICNA0188247501
Ecolite(+)™ liquid scintillation cocktail, MP Biomedicals
Szintillationsgemische
EcoLite(+)™ is a unique formulation of media possessing exceptional water and salt holding characteristics. Ready-to-use scintillation cocktail for a wide variety of aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is essential.
- Environmentally safe
- Non toxic and non flammable
- Ready-to-use
- Versatile
- Excellent stability for RIA and receptor assays
- Forms no gel or phasing
- Ideal for all types of samples
Suitable for difficult-to-count RIA samples, basic samples, phosphate buffer samples, and biological fluids.