AIN-76 vitamin mixture, MP Biomedicals
Lieferant: MP Biomedicals
0290545401
ICNA0290545401EA 225 EUR
ICNA0290545401
AIN-76 vitamin mixture, MP Biomedicals
Tierbedarf Tiernahrung
AIN-76 vitamin mixture is a basic component of animal diet for mice and rats colonies in the research lab.
Please see product datasheet for complete diet composition.
Verpackung: In powdered form.