Vitamin mixture, AIN-93-VX, MP Biomedicals
Lieferant: MP Biomedicals
0296040201
ICNA0296040201EA 225 EUR
ICNA0296040201
Vitamin mixture, AIN-93-VX, MP Biomedicals
Tierbedarf Tiernahrung
AIN-93-VX vitamin mixture is a component of AIN-93M and AIN-93G diet.
This vitamin mix is used for the rodents diet revised after numerous nutritional and technical problems were encountered, especially during growth, pregnancy, and lactation.
Bitte beachten Not available in pellet form.