Natriumpyruvat 0.1 mol/l in wässriger Lösung, steril

Lieferant: MP Biomedicals

Synonyme: 2-Oxopropionsäure Natriumsalz, Brenztraubensäure Natriumsalz, 2-Ketopropionsäure Natriumsalz, α-Ketopropionsäure Natriumsalz

091682049
ICNA091682049EA 19.2 EUR
ICNA091682049
Natriumpyruvat 0.1 mol/l in wässriger Lösung, steril
Natriumpyruvat

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Kreb′s cycle. The 100 mM solution should be diluted 1:100 for most cell culture. The use of sodium pyruvate in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described. A protocol that uses sodium pyruvate to establish stably transfected human B cell lines has been published. It improves coliform recovery when present in culture medium.


  • Application: Primary Cell Culture
  • pH: 6,8 (Lit.)
  • Osmolality: 430 to 500 mOsm/kg H₂O


In muscle, pyruvic acid (derived from glycogen) is reduced to lactic acid during exertion, which is reoxidised and partially retransformed to glycogen during rest. Involved in a metabolic regulatory pathway activated by mitochondrial oxidants. Pyruvate is involved in respiratory regulation in plants by interacting with alternative oxidase at a conserved cysteine residue. May help prevent hydrogen peroxide mediated cell death. Pyruvate, the anion of pyruvic acid, is the end product of the glycolysis pathway, whereby glucose is converted to pyruvate with the production of ATP.

Formel: CH₃COCOONa
Molecular Weight: 110,04 g/mol
Dichte: ~1,267 g/ml (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002586
CAS-Nummer: 113-24-6
EINECS: 204-024-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR