Cryopreservation media, Cellvation, MP Biomedicals
Lieferant: MP Biomedicals
Cellvation is a cryopreservation medium designed for optimal cell recovery and convenience.
- Optimal cell recovery and convenience
- No DMSO or serum
- Animal component free
- Preserve stem cells, hepatocytes, and tissue samples.
Cellvation does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as being ideally suited for all types of serum. Cellvation cryopreservation media is animal-component-free and has been shown to preserve the potential of stem cells for differentiation. In addition, the medium is recommended for the preservation of hepatocytes, tissue samples, and a wide variety of cell types and established cell lines.