Glass beads, MP Biomedicals
Lieferant: MP Biomedicals
116914801
ICNA116914801EA 107 EUR
ICNA116914801
Glass beads, MP Biomedicals
Kugeln Glaskugeln
Glass beads.
  • For research use only
  • Can be stored at 2 to 8 °C
Glass beads used as packing for distillation columns, mixing beads and boiling stones.