Digital Microscope
Lieferant: Dino-Lite / IDCP B.V.
AM4117MT-G2FBW
IDCPAM4117MT-G2FBWEA 1099 EUR
IDCPAM4117MT-G2FBW
Digital Microscope
Mikroskope
With 465 nm excitation light and a 510 - 545 nm emission bandpass filter range, AM4117MT-G2FBW is the ideal solution for the selective isolation and observation of samples with green fluorescence.
  • Green fluorescence
  • High sensitivity sensor
  • Adjustable LED intensity
  • Robust metallic housing
  • Interchangeable caps
The interchangeable caps provide adaptability to numerous applications with alternative lighting or object interface. The interchangeable front caps are designed to be used with Dino-Lite Edge and EdgePLUS series models. The caps provide additional versatility to Dino-Lite digital microscope applications.
The included caps may vary per model.
Provides 2 years warranty.
Verpackung: Microscope, carry pouch, software CD, calibration target, user manual.