Microscope clamp, gooseneck, Dino-Lite RK-02
Lieferant: Dino-Lite / IDCP B.V.
RK-02
IDCPRK-02EA 149 EUR
IDCPRK-02
Microscope clamp, gooseneck, Dino-Lite RK-02
Klemmen Klemmen für den allgemeinen Gebrauch
The RK-02 is a durable gooseneck clamp designed for flexible and versatile usage with any Dino-Lite digital microscope.
  • C-clamp fits to virtually any surface up to 7 cm (~3 inch) thick
  • Can be mounted horizontally or vertically
With this sturdy stand, users can easily position the Dino-Lite by bending the 60 cm long arm and adjusting the rotating head. The base of the RK-02 can also be rotated providing even more freedom and it can be fixed with the quick release fastener for use on uneven surfaces.
Compatible with all Dino-Lite models, except the high magnification models.