Manual Vacuum Regulation Turret with On and Off, Flush Mount
Lieferant: Welch by Gardner Denver
700563
ILMV700563EA 315.5 EUR
ILMV700563
Manual Vacuum Regulation Turret with On and Off, Flush Mount
Ventile Vakuum-Ventile
Quick opening/closing via ball valve of vacuum line plus manual vacuum control.
  • Dimensions: 690 mm
  • Diameter: 820 mm protrusion from wall and 20 mm into wall
Stainless steel On/Off ball valve allows easy repeat of flow rate setting.
Connect PTFE 10 mm ext.Ø tubing to valve using a PVDF compression fitting. SS Ball Valve has stepped hose barb that accepts 6,35 to 9,52 mm vacuum hose (DN 8-10).