High resolution colony counter, Scan® 1200
Lieferant: INTERSCIENCE
437000
INSC437000EA 0 EUR
INSC437000
High resolution colony counter, Scan® 1200
Kolonienzähler
High resolution colony counter that can be linked to a PC via a FireWire connection.
  • Minimum size of colonies is 0,05 mm
  • Detects up to seven colours on the same Petri dish
  • Measurement of the inhibition zone
Can count colonies in less than one second for a fast, accurate, and traceable reading of the result.