Gentamicinsulfat, ultrarein

Lieferant: Thermo Scientific
Gefahr

Synonyme: Gentamicin Sulfatsalz

J16051.14
J16051.14EA 0 EUR
J16051.14
Gentamicinsulfat, ultrarein
Gentamicinsulfat

  • Very stable
  • Autoclavable
  • Requires no refrigeration


Effective when combined with an antibiotic that interferes with cell wall synthesis.

Schmelzpunkt: 218…237 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270181
CAS-Nummer: 1405-41-0
Merck Index: 14,04392

Bestellen

Spezifikation Testergebnisse

Assay (Unspecified) - Activity Dry Basis ≥590 µg of gentamicin/mg
Assay (unspecified) Gentamicin Content (C1 + C2b) 25 - 50%
Assay (unspecified) Gentamicin Content (C1a) 10 -35%
Assay (unspecified) Gentamicin Content (C2 + C2a) 25 - 55%
Identity (A, B) Passes Test
Residue on Ignition ≤1.0%
Impurity Content Limit of Methanol ≤1.0%
Loss on Drying ≤18.0%
Optical Rotation (c=1 in H₂O) +107 to 121°
pH (4%, H₂O) 3.5 - 5.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR