Glycin (H-Gly-OH) ≥99,5% Reagenz für die Zellkultur
Lieferant: Thermo Scientific
Synonyme:
Aminoacetic acid, Glycocoll, H-Gly-OH
J62407.A1
J62407.A1EA
69.5
EUR
J62407.A1
Glycin (H-Gly-OH) ≥99,5% Reagenz für die Zellkultur
Glycine
Purity: 99.5+%
CAS No.: 56-40-6
Molecular Formula: C2H5NO2
Formula Weight: 75.07
Physical Form: Powder
Density: 1.595
Appearance: White to off-white
Odour: Odourless
pH: 5,97
Melting Point: ca 245° dec.
Beilstein Reference: 635782
Merck Reference: 14,4491
MDL No.: MFCD00008131
Formel:
NH₂CH₂COOH Molecular Weight: 75,07 g/mol Siedepunkt: 181 °C (1013 hPa) Schmelzpunkt: ∼ 245 °C (Zersetzung) Dichte: 1,595 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008131 CAS-Nummer: 56-40-6 EINECS: 200-272-2 Merck Index: 14,4491 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...