Quick release, 4-point Linesman chinstrap harness
Lieferant: JSP
AHV200-000-000
JSPAAHV200-000-000EA 16.5 EUR
JSPAAHV200-000-000
Quick release, 4-point Linesman chinstrap harness
Schutzhelme Schutzhelme
The JSP 4-point Linesman chinstrap provides increased security for a firmer fit, preventing helmets being accidentally dropped during extreme movement.
  • 4-point chinstrap for Evolution® safety helmets
  • Tubular nylon webbing
  • LDPE clip
  • Nylon buckle
  • Increased security for a firmer fit
The JSP 4-point Linesman chinstrap provides increased security for a firmer fit, preventing helmets from being accidentally dropped during extreme movement. Simple, secure closure from the nylon buckle allows quick donning and doffing of the helmet.
Standards: EN 397 EN 14052. Meets the requirements of EN 397 and EN 14052 when worn with the appropriate JSP EVO® helmet.