7000 Safety spectacle, Junior Stealth™
Lieferant: JSP
ASA918-321-100
JSPAASA918-321-100EA 5.6 EUR
JSPAASA918-321-100
7000 Safety spectacle, Junior Stealth™
Schutzbrillen
Contour design and flexibility of frame give a good versatile fit onto both smaller and larger faces.
  • One-piece lens
  • Permanently attached side arms for durability
  • Optical class 1
The one-piece lens features permanently attached side arms for durability. Lens and frame offer protection against low energy, high-speed particles at 45 m/s (101 mph). The optical qualities of the lens comply with the requirements of optical class 1 (highest).
Standards: Conforms to EN 166.