Digital temperature controller, LTR 2500
Lieferant: JUCHHEIM SOLINGEN
43900118
JUCE43900118EA 639 EUR
JUCE43900118
Digital temperature controller, LTR 2500
Regler Temperaturregler
Optimised temperature controller with plastic housing.
  • Actual value permanently visible, nominal value set via pushbutton
  • Overtemperature switch-off function at nominal value
  • Security controller with sensor input and shut-down
Suitable for direct connection of laboratory heaters, resistance thermometers, and thermocouples.
Nominal contant:+>10K