VWR® A.C.E.™, Sequencing buffers
Lieferant: VWR Chemicals
A.C.E.™ Sequencing Buffers are optimised buffers for DNA capillary electrophoresis sequencing that provide excellent results like long read lengths, high resolution, and consistent run times.
- Compatible with ABI Prism 3730, 3700, 3100, or 310 and the Beckman CEQ 2000/CEQ 8000 automated sequencers
- Long read lengths
- High resolution
- Consistent run times
- Storage at room temperature
The tinted sequencing buffers have a slight pink hue in order to help the user avoid confusion. Once loaded, the researcher will be able to differentiate between A.C.E.™ buffer and any other water, buffer, or solution in the sequencer. Tinted A.C.E.™ buffers have the same high-quality formulation delivering consistent and reproducible results.
This 10X sequencing buffer concentrate is conveniently diluted to the desired working concentration, thereby providing an easy and cost-effective buffer for the operating systems and 1X sequencing buffer comes ready-to-use and no dilution required, offering convenience and time savings.