Scheidetrichter, birnenförmig
Lieferant: Kavalier
632426355940
KAVA632426355940EA 367 EUR
KAVA632426355940
Scheidetrichter, birnenförmig
Trichter Separating Funnels
Made of borosilicate glass.
  • With stopper
  • Stopcock with glass key
  • Very good chemical and temperature resistance