Automatische Pipetten, Kipp, mit Flasche
Lieferant: Kavalier
632446003005
KAVA632446003005EA 3490 EUR
KAVA632446003005
Automatische Pipetten, Kipp, mit Flasche
Kippautomaten
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head with standard ground jont NS 29/32 is completed by a 250 ml flask for repetitive dispensing of the same volume.