Dual stage pressure regulators
Lieferant: KAYSER WERK
CK1310
KAYSCK1310EA 379 EUR
KAYSCK1310
Dual stage pressure regulators
Druckminderer
The set outlet pressure is kept constant regardless of the bottle level.
Standards: Manufactured according to EN ISO 2503.