Reaction vessels, cylindrical, with flat bottom and thermal jacket
Lieferant: KGW
TRGN7399
KGWATRGN7399EA 0 EUR
KGWATRGN7399
Reaction vessels, cylindrical, with flat bottom and thermal jacket
Reaktionsgefäße Reaktionsgefäße, zylindrisch
This cylindrical reaction vessel is made up of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
It has flat bottom and thermal jacket, GL 18 Olive tubing connection.