Surface Cleanser, Abrasive Liquid
Lieferant: KING DISPOSABLES
PS10161
KINGPS10161EA 41.1 EUR
KINGPS10161
Surface Cleanser, Abrasive Liquid
Reinigungsmittel Waschmittel und Oberflächenreiniger
Cleaner for all ceramic, glased, stainless steel surfaces (excluding brushed stainless steel) in sanitary areas and in the kitchen.