Utility washer
Lieferant: KLEINFELD
459960-0000
KLEI459960-0000EA 537 EUR
KLEI459960-0000
Utility washer
Reinigungs- und Desinfektionsautomaten
This washer is used for rapid, safe washing and drying of cuvettes, tubes and small scale volumetric items up to approximately 22 mm OD.
  • Operates efficiently with a conventional sink aspirator
  • Recommended for 10 mm NMR tubes
  • Flushing is activated by lightly pressing the inverted cell onto a neoprene gasket
All glass apparatus manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Lieferumfang: Supplied with one gasket, a tube washer, an Erlenmeyer flask and a ³/₈" hose connection.