pH electrode, SE 102-MS
Lieferant: KNICK
SE102-MS
KNICSE102-MSEA 361 EUR
KNICSE102-MS
pH electrode, SE 102-MS
Elektroden pH-Elektroden
This pH sensor with high reliability and precision, digital, with Memosens technology for perfect galvanic isolation. The special pH glass enables a very fast and stable response and makes the sensor to use in broad temperature range, even in the harshest of environments.
  • No influence of humidity in the connector
  • Liquid electrolyte, refillable and this can be pre calibrated
  • Digital data transfer
  • Integrated sensor diagnostics
  • Integrated temperature detector NTC 30 kΩ
  • Ceramic junction
Special pH glass with low impedance; suitable for measurements in a broad temperature range, including low temperatures.
The liquid electrolyte of the reference electrode can be refilled. Designed for a wide range of applications, the sensor is ideal for measuring with Portavo portable analysers.
Applied in chemistry, quality control, food and beverages, aggressive media.