Precision pliers, electronic super Knips®
Lieferant: KNIPEX WERKE
7803125
KNIP7803125EA 38.3 EUR
KNIP7803125
Precision pliers, electronic super Knips®
Zangen
Precision pliers suitable for ultra fine cutting work.
  • Ground, very sharp cutting edges without bevel
  • Precision shaped tips cut wires resting on a board from 0,2 mm diameter
  • Joint with stainless steel rivet
  • Extremely smooth movement for minimum operator fatigue
  • With opening spring and opening limiter
Shear cut with controlled micro cutting edge misalignment for the most precise cutting of even the thinnest of wires and for a long service life.
Cutting capacities for soft wire (0,2 to 1,6 mm) and medium hard wire 1,0 mm diameter.
Standards: DIN ISO 9654.