Iron(II) ethylenediammonium sulphate tetrahydrate , Basic substance rückführbar auf NIST-Standardreferenzmaterial (SRM)
Lieferant: Bernd Kraft
Synonyme:
Ferrous Ethylenediamine Sulphate
20480.0601
KRAF20480.0601EA
570
EUR
KRAF20480.0601
Iron(II) ethylenediammonium sulphate tetrahydrate , Basic substance rückführbar auf NIST-Standardreferenzmaterial (SRM)
Iron(II) ethylenediammonium sulphate tetrahydrate
Formel:
C₂H₈N₂ Molecular Weight: 310,07 g/mol |
MDL:
MFCD00082474 CAS-Nummer: 113193-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...