Handheld turbidity meter
Lieferant: LaMotte Chemicals
LMI974-T
LAMOLMI974-TEA 3270 EUR
LAMOLMI974-T
Handheld turbidity meter
Trübungsmessgeräte
Portable turbidity meter for both low level drinking water applications and high turbidity.
  • Large six-line backlit LCD
  • Lithium rechargeable battery
  • Seven user selectable languages: English, French, Spanish, Japanese, Italian, Chinese, Portuguese
Lieferumfang: Supplied with 0,1 and 10 NTU standards, sample bottle, four sample tubes, USB cable, power adapter (USB) and carrying case.