Adjustable Height Base Stands
Lieferant: LABCONCO
3780401
LBCP3780401EA 0 EUR
LBCP3780401
Adjustable Height Base Stands
Dunstabzugshauben und Verkleidungen Chemikalien-Abzüge
For use with Basic™ 47, Basic 70, Fiberglass 30, Protector® XStream®, Protector Premier™, Protector ClassMate®, Protector PVC perchloric acid, protector stainless steel perchloric acid, logic+, logic, cell logic, puriCare procedure stations and protector radioisotope laboratory hoods.
  • Telescoping base stands with fixed feet can be adjusted to eight height positions prior to installation
  • Telescoping legs are adjustable in 25,4 mm increments from 698,5 to 876,3 mm
  • Durable 44,45 mm tubular, epoxy-coated steel frame
Base stands are constructed of durable 45 mm tubular, glacier white epoxy-coated steel and support loads up to 453,6 kg. ADA-compliant. Stands with telescoping legs are adjustable from 699 to 851 mm high. Hydraulic lift base stands adjust from 648 to 851 mm high and include four powder-coated, die-cast feet with levelers. Manual hydraulic lift models include front-accessible hand crank. Electric hydraulic lift models include a front-accessible up/down switch.
Standards: Electrical components of hydraulic lift stands are UL listed.
Bestellinformation: Hoods can be placed on a base stand or base cabinet. Several base cabinet models are available: standard storage (see 46610-404 series), acid storage (see 46610-412 series), and solvent storage (see 46610-440 series).