Distilling bridges acc. to Claisen
Lieferant: Lenz Laborglas GmbH & CO.KG
05055314
LENZ05055314EA 112 EUR
LENZ05055314
Distilling bridges acc. to Claisen
Destillationsbauteile Destillierbrücken
Still heads with Liebig condenser and Claisen attachment made of DURAN® borosilicate glass. Straight outlet. Socket for distillation thermometer with 55 mm total length. Liebig condenser with straight outlet. Connection for coolant water via DIN threated GL 14 connectors.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes