Contact plates (Rodac) in peelable blister pack
Lieferant: LIOFILCHEM
15325
15327
15328
15329
15331
15332
15362
15334
15365
15376
15360
15205
15389
LIOF15325EA
62.1
EUR
LIOF15325
LIOF15327
LIOF15328
LIOF15329
LIOF15331
LIOF15332
LIOF15362
LIOF15334
LIOF15365
LIOF15376
LIOF15360
LIOF15205
LIOF15389
Contact plates (Rodac) in peelable blister pack
Medien für die Mikrobiologie
Abklatschplatten für die Mikrobiologie
The new peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
- Allows the usage of single plates, leaving the remaining plates protected in their pack
- Drastically reduces the dehydration of culture medium during the shelf life period
- Permits better product preservation and isolation during transportation and storage
- Guarantees the product cannot be handled after the packaging
- Avoids contamination of the controlled environment