Contact Plates, Blister
Lieferant: LIOFILCHEM
15339S 15323S 15326 15327S 15336 15340S 15375 15380S 15336S 15203S 15204S 15361
LIOF15339SEA 64.3 EUR
LIOF15339S LIOF15323S LIOF15326 LIOF15327S LIOF15336 LIOF15340S LIOF15375 LIOF15380S LIOF15336S LIOF15203S LIOF15204S LIOF15361
Contact Plates, Blister
Medien für die Mikrobiologie Abklatschplatten für die Mikrobiologie
The peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
  • Full transparency of the package
  • No release particles during the film opening phase, critical in environment controlled by particle counters
  • Allows the usage of single plates, leaving the remaining plates protected in their pack
  • Drastically reduces the dehydration of culture medium during the shelf life period
  • Permits better product preservation and isolation during transportation and storage
  • Guarantees the product cannot be handled after the packaging and avoids contamination