Contact Plates, Blister
Lieferant: LIOFILCHEM
15339S
15323S
15326
15327S
15336
15340S
15375
15380S
15336S
15203S
15204S
15361
LIOF15339SEA
64.3
EUR
LIOF15339S
LIOF15323S
LIOF15326
LIOF15327S
LIOF15336
LIOF15340S
LIOF15375
LIOF15380S
LIOF15336S
LIOF15203S
LIOF15204S
LIOF15361
Contact Plates, Blister
Medien für die Mikrobiologie
Abklatschplatten für die Mikrobiologie
The peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
- Full transparency of the package
- No release particles during the film opening phase, critical in environment controlled by particle counters
- Allows the usage of single plates, leaving the remaining plates protected in their pack
- Drastically reduces the dehydration of culture medium during the shelf life period
- Permits better product preservation and isolation during transportation and storage
- Guarantees the product cannot be handled after the packaging and avoids contamination