Latex tests
Lieferant: LIOFILCHEM
96147
LIOF96147EA
219
EUR
LIOF96147
Latex tests
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Latex
Identification of the main pathogenic microorganisms directly from clinical specimens.
- Save time and labor with direct testing
- Results in 18 to 24 hours
- Confirmation with latex agglutination tests
- Clear and well defined colour changes
- Easy interpretation